Gene Transcripts

Gene Symbol
Vps35
Entrez Gene ID
65114
Gene Name
vacuolar protein sorting 35
Gene Aliases
AI647796, Mem3
Chromosome Location
Chr. 8: 85260392 - 85299497 on Build GRCm38
UniGene ID
Mm.296520
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022997.4 NP_075373.1 8 901
GenBank mRNA AF226323.1 - 8 920
AK135680.1 - 8 1207
AK142382.1 - 8 1188
AK145421.1 - 7 869
AK145853.1 - 8 950
AK150682.1 - 8 915
AK162726.1 - 8 880
AK166821.1 - 8 894
AK167421.1 - 8 918
AK167594.1 - 8 914
AK168880.1 - 8 894
AK170473.1 - 8 935
AK218937.1 - 2 153
BC005469.1 - 8 926 3492842
BC006637.1 - 8 906 3969440
U47024.1 - 7 869

View Full Product Details