Gene Transcripts

Gene Symbol
Fam213b
Entrez Gene ID
66469
Gene Name
family with sequence similarity 213, member B
Gene Aliases
2810405K02Rik, AI836168, PM/PGFS
Chromosome Location
Chr. 4: 154896430 - 154899043 on Build GRCm38
UniGene ID
Mm.41868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025582.3 NP_079858.2 7 765
GenBank mRNA AK005188.1 - 7 766
AK012999.1 - 7 764
BC030453.1 - 7 759 5401948

View Full Product Details