Gene Transcripts

Gene Symbol
Zfp830
Entrez Gene ID
66983
Gene Name
zinc finger protein 830
Gene Aliases
2410003C20Rik, AV301118, AW048207, Ccdc16, Omcg1, Znf830
Chromosome Location
Chr. 11: 82764345 - 82767622 on Build GRCm38
UniGene ID
Mm.29622
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025884.4 NP_080160.2 1 613
GenBank mRNA AK008475.1 - 1 613
AK010353.1 - 1 604
AK017640.1 - 1 593
AK050043.1 - 1 608
AK132497.1 - 1 614
AK148058.1 - 1 614
AK167656.1 - 1 604
AK168417.1 - 1 593
BC024340.1 - 1 599 4981533

View Full Product Details