Gene Transcripts

Gene Symbol
Zfp830
Entrez Gene ID
66983
Gene Name
zinc finger protein 830
Gene Aliases
2410003C20Rik, AV301118, AW048207, Ccdc16, Omcg1, Znf830
Chromosome Location
Chr. 11: 82764345 - 82767622 on Build GRCm38
UniGene ID
Mm.29622
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025884.4 NP_080160.2 1 1305
GenBank mRNA AK008475.1 - 1 1305
AK010353.1 - 1 1296
AK050043.1 - 1 1300
AK132497.1 - 1 1306
AK148058.1 - 1 1306
AK163064.1 - 1 368
AK167656.1 - 1 1296
AK168417.1 - 1 1285
BC024340.1 - 1 1291 4981533

View Full Product Details