Gene Transcripts

Gene Symbol
STAG2
Entrez Gene ID
10735
Gene Name
stromal antigen 2
Gene Aliases
SA-2, SA2, SCC3B, bA517O1.1
Chromosome Location
Chr. X: 123960560 - 124102656 on Build GRCh38
UniGene ID
Hs.496710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042749.2 NP_001036214.1 35 4334
NM_001042750.1 NP_001036215.1 35 4178
NM_001042751.1 NP_001036216.1 34 4067
NM_001282418.1 NP_001269347.1 34 4046
NM_006603.4 NP_006594.3 33 4002
XM_005262357.2 XP_005262414.1 34 4203
XM_005262358.2 XP_005262415.1 35 4140
XM_005262359.3 XP_005262416.1 34 4093
XM_005262360.2 XP_005262417.1 34 4022
XM_005262361.2 XP_005262418.1 33 4039
XM_006724727.1 XP_006724790.1 36 4282
XM_006724728.3 XP_006724791.1 35 4216
XM_011531253.2 XP_011529555.1 34 4075
XM_017029232.1 XP_016884721.1 34 4029
XM_017029233.1 XP_016884722.1 33 3911
XM_017029234.1 XP_016884723.1 33 3964
XM_017029235.1 XP_016884724.1 35 4221
GenBank mRNA AK026678.1 - 1 43
AK298633.1 - 23 2721
AL831939.2 - 33 3970
BC001765.1 - 33 3951 3352347
BX640970.1 - 35 4189
BX641002.1 - 34 3971
BX641003.1 - 34 4036
CR627425.1 - 34 4045
CR749433.1 - 35 4147

View Full Product Details