Gene Transcripts

Gene Symbol
Sf3b5
Entrez Gene ID
66125
Gene Name
splicing factor 3b, subunit 5
Gene Aliases
10kDa, 1110005L13Rik, AU043053, Sf3b10
Chromosome Location
Chr. 10: 13008450 - 13009183 on Build GRCm38
UniGene ID
Mm.29743
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175102.4 NP_780311.2 1 647
GenBank mRNA AK003475.1 - 1 616
AK009874.1 - 1 610
AK010348.1 - 1 622

View Full Product Details