Gene Transcripts
- Gene Symbol
- Slc39a1
- Entrez Gene ID
- 30791
- Gene Name
- solute carrier family 39 (zinc transporter), member 1
- Gene Aliases
- Zip1, Zirtl
- Chromosome Location
- Chr. 3: 90248192 - 90253613 on Build GRCm38
- UniGene ID
- Mm.294709
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC005474.1 |
- |
4 |
540 |
|
View Full Product Details