Gene Transcripts

Gene Symbol
Slc39a1
Entrez Gene ID
30791
Gene Name
solute carrier family 39 (zinc transporter), member 1
Gene Aliases
Zip1, Zirtl
Chromosome Location
Chr. 3: 90248192 - 90253613 on Build GRCm38
UniGene ID
Mm.294709
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013901.2 NP_038929.2 4 1624
GenBank mRNA AK159290.1 - 4 1481
BC005474.1 - 4 1586 3591856
BC138459.1 - 3 1561 8861479

View Full Product Details