Gene Transcripts

Gene Symbol
Rgs6
Entrez Gene ID
50779
Gene Name
regulator of G-protein signaling 6
Gene Aliases
Chromosome Location
Chr. 12: 82616176 - 83158815 on Build GRCm38
UniGene ID
Mm.153013
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282061.1 NP_001268990.1 5 410
NM_015812.3 NP_056627.1 5 692
GenBank mRNA AF061933.1 - 2 137
AK052527.1 - 5 693
AK053087.1 - 5 411
AK139323.1 - 5 807
BC038616.1 - 3 237
BC125029.1 - 5 396 40062355
BC125030.1 - 5 396 40062356

View Full Product Details