Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016767.2 | NP_058047.1 | 3 | 514 | |
GenBank mRNA | AF017021.1 | - | 3 | 514 | |
AK018587.1 | - | 3 | 556 | ||
BC132410.1 | - | 3 | 348 | 40130687 | |
BC132412.1 | - | 3 | 348 | 40130689 |