Gene Transcripts

Gene Symbol
Snx3
Entrez Gene ID
54198
Gene Name
sorting nexin 3
Gene Aliases
SDP3
Chromosome Location
Chr. 10: 42502054 - 42535369 on Build GRCm38
UniGene ID
Mm.277870
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017472.4 NP_059500.2 4 803
GenBank mRNA AF062482.1 - 4 601
AK007798.1 - 4 614
AK013183.1 - 4 828
AK013261.1 - 4 436
AK018790.1 - 4 436
AK145350.1 - 4 606
AK154728.1 - 4 606
BC116885.1 - 4 647 40126204
BC116889.1 - 4 647 40126208

View Full Product Details