Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019455.4 | NP_062328.3 | 2 | 184 | |
GenBank mRNA | AK020246.1 | - | 2 | 185 | |
AK039392.1 | - | 2 | 185 | ||
BC116735.1 | - | 2 | 152 | 40126054 | |
BC116737.1 | - | 2 | 152 | 40126056 | |
D82072.1 | - | 2 | 362 |