Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019480.4 | NP_062353.3 | 5 | 699 | |
GenBank mRNA | AF076524.1 | - | 4 | 382 | |
AK004366.1 | - | 4 | 407 | ||
AK159499.1 | - | 5 | 749 | ||
AK160149.1 | - | 5 | 868 | ||
AY009091.1 | - | 5 | 703 | ||
BC026717.1 | - | 5 | 454 | ||
BC108397.1 | - | 5 | 699 | 6329278 | |
BC119118.1 | - | 5 | 615 | 8733867 | |
BC119120.1 | - | 5 | 615 | 8733869 | |
BC144740.1 | - | 5 | 649 | 9053288 |