Gene Transcripts

Gene Symbol
Gsk3b
Entrez Gene ID
56637
Gene Name
glycogen synthase kinase 3 beta
Gene Aliases
7330414F15Rik, 8430431H08Rik, C86142, GSK-3, GSK-3beta, GSK3
Chromosome Location
Chr. 16: 38089001 - 38246080 on Build GRCm38
UniGene ID
Mm.394930
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019827.6 NP_062801.1 3 1842
GenBank mRNA AB066114.1 - 3 1841
AF156099.2 - 3 483
AK154293.1 - 3 1320
AK164048.1 - 2 194
AK170965.1 - 3 1289
AK190946.1 - 3 286
BC006936.1 - 3 349 2648507
BC060743.1 - 3 1769 4022374

View Full Product Details