Gene Transcripts

Gene Symbol
Nudt3
Entrez Gene ID
56409
Gene Name
nudix (nucleotide diphosphate linked moiety X)-type motif 3
Gene Aliases
1110011B09Rik, AA960325, Dipp, Dipp1
Chromosome Location
Chr. 17: 27579382 - 27623452 on Build GRCm38
UniGene ID
Mm.144699
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291046.1 NP_001277975.1 5 1281
NM_019837.2 NP_062811.1 5 1273
GenBank mRNA AF264064.1 - 5 1273
AK003601.1 - 1 264
AK079658.1 - 3 769
BC016534.1 - 6 1180 2615904
BC046805.1 - 5 1273 6829170

View Full Product Details