Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019917.2 | NP_064301.2 | 2 | 248 | |
GenBank mRNA | AF053985.1 | - | 2 | 248 | |
AY426342.1 | - | 2 | 248 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001104641.1 | NP_001098111.1 | 2 | 248 |