Gene Transcripts
- Gene Symbol
- Stap1
- Entrez Gene ID
- 56792
- Gene Name
- signal transducing adaptor family member 1
- Gene Aliases
- AI586015, Brdg1, STAP-1
- Chromosome Location
- Chr. 5: 86071585 - 86104784 on Build GRCm38
- UniGene ID
- Mm.131237
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019992.3 |
NP_064376.1
|
1, 2 |
135 |
|
GenBank mRNA |
AB036058.1 |
- |
1, 2 |
125 |
|
AK041474.1 |
- |
1, 2 |
136 |
|
AK042475.1 |
- |
1, 2 |
182 |
|
AK153661.1 |
- |
1, 2 |
214 |
|
AK162655.1 |
- |
1, 2 |
217 |
|
BC057898.1 |
- |
1, 2 |
189 |
|
BC145550.1 |
- |
1, 2 |
151 |
9054114 |
BC145828.1 |
- |
1, 2 |
150 |
40131131 |
BC145830.1 |
- |
1, 2 |
151 |
40131133 |
View Full Product Details