Gene Transcripts

Gene Symbol
Mocs1
Entrez Gene ID
56738
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
3110045D15Rik, AI043170, AW536397
Chromosome Location
Chr. 17: 49428364 - 49455435 on Build GRCm38
UniGene ID
Mm.22256
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020042.2 NP_064426.2 11 2587
NM_028464.1 NP_082740.1 11 2602
GenBank mRNA AK157231.1 - 11 2537
BC027444.1 - 3 1507
BC034066.1 - 7 1945
BC049914.1 - 9 2224
BE851067.1 - 1 473

View Full Product Details