Gene Transcripts

Gene Symbol
Tslp
Entrez Gene ID
53603
Gene Name
thymic stromal lymphopoietin
Gene Aliases
Chromosome Location
Chr. 18: 32815383 - 32819799 on Build GRCm38
UniGene ID
Mm.143716
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021367.2 NP_067342.1 5 1009
GenBank mRNA AF232937.1 - 5 1008
AK087266.1 - 2 1013
CJ198416.1 - 1 239

View Full Product Details