Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083118.2 | NP_001076587.1 | 6 | 906 | |
NM_001286200.1 | NP_001273129.1 | 6 | 906 | ||
NM_009353.2 | NP_033379.2 | 6 | 906 | ||
GenBank mRNA | AF003000.1 | - | 6 | 800 | |
AK042508.1 | - | 1 | 427 | ||
AK049507.1 | - | 6 | 906 | ||
AK089419.1 | - | 6 | 881 | ||
AK160421.1 | - | 7 | 651 | ||
BC046284.1 | - | 6 | 895 | 6439629 |