Gene Transcripts

Gene Symbol
Vmn2r10
Entrez Gene ID
22307
Gene Name
vomeronasal 2, receptor 10
Gene Aliases
V2r16, VR16
Chromosome Location
Chr. 5: 108995539 - 109006436 on Build GRCm38
UniGene ID
Mm.261598
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009491.3 NP_033517.3 1 174
GenBank mRNA AF011426.1 - 209

View Full Product Details