Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006810.3 | NP_006801.1 | 17 | 1712 | |
NR_028444.1 | 16 | 1556 | |||
GenBank mRNA | AK054963.1 | - | 5 | 2084 | |
BC001625.1 | - | 16 | 1503 | 3537659 | |
D49490.1 | - | 17 | 1612 |