Gene Transcripts

Gene Symbol
Clcn6
Entrez Gene ID
26372
Gene Name
chloride channel 6
Gene Aliases
AI850629
Chromosome Location
Chr. 4: 148004259 - 148038858 on Build GRCm38
UniGene ID
Mm.89987
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011929.2 NP_036059.1 16 1615
GenBank mRNA AK145053.1 - 16 1660
AK165362.1 - 16 1665
AK220319.1 - 10 1105

View Full Product Details