Gene Transcripts

Gene Symbol
Trpc7
Entrez Gene ID
26946
Gene Name
transient receptor potential cation channel, subfamily C, member 7
Gene Aliases
TRP7, Trrp8
Chromosome Location
Chr. 13: 56773098 - 56895974 on Build GRCm38
UniGene ID
Mm.62409
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF139923.1 - 3 967
AK135153.1 - 3 1156
AK137418.1 - 3 1168
BC117995.1 - 3 967 40096022

View Full Product Details