Gene Transcripts

Gene Symbol
Cops5
Entrez Gene ID
26754
Gene Name
COP9 (constitutive photomorphogenic) homolog, subunit 5 (Arabidopsis thaliana)
Gene Aliases
AI303502, CSN5, Jab1, Mov34, Sgn5
Chromosome Location
Chr. 1: 10024600 - 10038159 on Build GRCm38
UniGene ID
Mm.402384
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013715.2 NP_038743.1 1, 2 485
GenBank mRNA AF065386.1 - 1, 2 238
AF068223.1 - 1, 2 253
AK002761.1 - 1, 2 267
AK005728.1 - 1, 2 272
AK012499.1 - 1, 2 272
AK133073.1 - 1, 2 265
AK146271.1 - 1, 2 272
AK151492.1 - 1, 2 271
BC046753.1 - 1, 2 231 5693100
CX240457.1 - 1, 2 453
U70736.1 - 1, 2 261

View Full Product Details