Gene Transcripts

Gene Symbol
Pald1
Entrez Gene ID
27355
Gene Name
phosphatase domain containing, paladin 1
Gene Aliases
MMPAL, Pald, mKIAA1274
Chromosome Location
Chr. 10: 61319656 - 61383533 on Build GRCm38
UniGene ID
Mm.269699
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013753.2 NP_038781.2 20 4200
GenBank mRNA AK088577.1 - 20 4036
AK146489.1 - 20 4145
X99384.1 - 19 3986

View Full Product Details