Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013781.3 | NP_038809.1 | 2 | 317 | |
GenBank mRNA | AB043953.1 | - | 2 | 300 | |
AF168364.1 | - | 2 | 258 | ||
AK155165.1 | - | 2 | 253 | ||
AK172072.1 | - | 2 | 238 | ||
BC113203.1 | - | 2 | 272 | 40048064 |