Gene Transcripts

Gene Symbol
Zfp68
Entrez Gene ID
24135
Gene Name
zinc finger protein 68
Gene Aliases
KRAB3, KRAZ2, Zfp70, mszf22, mszf49
Chromosome Location
Chr. 5: 138603652 - 138619743 on Build GRCm38
UniGene ID
Mm.27575
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044747.2 NP_001038212.1 5 607
NM_001163797.1 NP_001157269.1 6 697
NM_013844.2 NP_038872.1 6 703
GenBank mRNA AB024005.1 - 6 687
AK013868.1 - 6 698
AK044831.1 - 5 577
AK146092.1 - 6 664
AK153887.1 - 6 639
BC018171.1 - 6 686 3989444
U29502.1 - 3 278

View Full Product Details