Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021885.4 | NP_068685.1 | 9 | 1251 | |
GenBank mRNA | AK147700.1 | - | 9 | 1306 | |
BC096554.1 | - | 9 | 1232 | 30616697 | |
U52433.1 | - | 9 | 1239 | ||
U52824.1 | - | 10 | 1384 | ||
U54643.1 | - | 9 | 1169 |