Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031884.1 | NP_114090.1 | 8 | 1014 | |
GenBank mRNA | AF312713.2 | - | 8 | 1102 | |
AK149569.1 | - | 8 | 985 | ||
AY195873.1 | - | 8 | 1102 | ||
BC106766.2 | - | 8 | 991 | 40039484 |