Gene Transcripts

Gene Symbol
Abcg5
Entrez Gene ID
114628
Gene Name
ATP-binding cassette, subfamily G (WHITE), member 5
Gene Aliases
Chromosome Location
Chr. 6: 7935771 - 7961339 on Build Rnor_6.0
UniGene ID
Rn.74258
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053754.2 NP_446206.2 2 317
XM_008764452.1 XP_008762674.1 2 449
GenBank mRNA AF312714.3 - 2 317

View Full Product Details