Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133528.1 | NP_598212.1 | 2, 3 | 135 | |
GenBank mRNA | AJ132008.1 | - | 2, 3 | 135 | |
BC085708.1 | - | 2, 3 | 156 | 7193103 | |
FQ229796.1 | - | 2, 3 | 151 | ||
FQ233766.1 | - | 2, 3 | 158 |