Gene Transcripts

Gene Symbol
Clec10a
Entrez Gene ID
64195
Gene Name
C-type lectin domain family 10, member A
Gene Aliases
Mgl, Mgl1, Mgll
Chromosome Location
Chr. 10: 56764892 - 56769447 on Build Rnor_6.0
UniGene ID
Rn.779
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022393.2 NP_071788.2 4 517
XM_006246803.2 XP_006246865.1 4 555
XM_008767867.1 XP_008766089.1 4 555
XM_008767868.1 XP_008766090.1 4 399
XM_008767869.1 XP_008766091.1 4 396
GenBank mRNA BC091151.1 - 4 517
FQ213042.1 - 4 389
FQ221204.1 - 4 406
FQ228437.1 - 4 375
J05495.1 - 4 377

View Full Product Details