Gene Transcripts
- Gene Symbol
- KPTN
- Entrez Gene ID
- 11133
- Gene Name
- kaptin, actin binding protein
- Gene Aliases
- 2E4, MRT41
- Chromosome Location
- Chr. 19: 47475141 - 47486792 on Build GRCh38
- UniGene ID
- Hs.25441
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291296.1 |
NP_001278225.1
|
8, 9 |
943 |
|
NM_007059.3 |
NP_008990.2
|
10, 11 |
1111 |
|
NR_111923.1 |
|
11, 12 |
1198 |
|
XM_011526399.2 |
XP_011524701.1
|
13, 14 |
2616 |
|
XM_017026226.1 |
XP_016881715.1
|
13, 14 |
1461 |
|
XM_017026227.1 |
XP_016881716.1
|
12, 13 |
1537 |
|
XM_017026228.1 |
XP_016881717.1
|
13, 14 |
1461 |
|
GenBank mRNA |
AF105369.1 |
- |
10, 11 |
1043 |
|
AK023996.1 |
- |
10, 11 |
1066 |
|
AK298672.1 |
- |
11, 12 |
1096 |
|
BC009249.2 |
- |
10, 11 |
1026 |
3029623 |
View Full Product Details