Gene Transcripts

Gene Symbol
Slc21a4
Entrez Gene ID
80899
Gene Name
kidney specific organic anion transporter
Gene Aliases
OAT-K1, OAT-K2, Slco1a3, rOAT-K1, rOAT-K11, rOAT-K13, rOAT-K14, rOAT-K2, rOAT-K3, rOAT-K5, rOAT-K6, rOAT-K7, rOAT-K8, rOAT-K9
Chromosome Location
Chr. 4: 176255603 - 176299119 on Build Rnor_6.0
UniGene ID
Rn.202967
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030837.1 NP_110464.1 16 2262
GenBank mRNA AB012662.1 - 15 2154
BC085730.1 - 9 1347 7110926
D79981.1 - 16 2262

View Full Product Details