Gene Transcripts
- Gene Symbol
- Chrm2
- Entrez Gene ID
- 81645
- Gene Name
- cholinergic receptor, muscarinic 2
- Gene Aliases
- Acm2
- Chromosome Location
- Chr. 4: 64089028 - 64091090 on Build Rnor_6.0
- UniGene ID
- Rn.10752
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031016.1 |
NP_112278.1
|
1 |
1449 |
|
GenBank mRNA |
J03025.1 |
- |
1 |
1449 |
|
View Full Product Details