Gene Transcripts

Gene Symbol
Adap1
Entrez Gene ID
171097
Gene Name
ArfGAP with dual PH domains 1
Gene Aliases
Centa1, p42IP4
Chromosome Location
Chr. 12: 17416327 - 17468212 on Build Rnor_6.0
UniGene ID
Rn.10539
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133567.2 NP_598251.1 2 257
GenBank mRNA AF123047.1 - 2 184
AJ007422.1 - 2 184
AJ007616.1 - 2 184
BC099775.1 - 2 257 7933317
U51013.1 - 2 284

View Full Product Details