Gene Transcripts

Gene Symbol
Sel1l
Entrez Gene ID
314352
Gene Name
sel-1 suppressor of lin-12-like (C. elegans)
Gene Aliases
Sel1h
Chromosome Location
Chr. 6: 115575135 - 115616857 on Build Rnor_6.0
UniGene ID
Rn.162892
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177933.2 NP_808794.2 5, 6 712
GenBank mRNA AF304853.1 - 5, 6 726
AF304855.1 - 1, 2 111
CB814385.1 - 2, 3 147

View Full Product Details