Gene Transcripts

Gene Symbol
Rtn4rl1
Entrez Gene ID
303311
Gene Name
reticulon 4 receptor-like 1
Gene Aliases
Ngrh2
Chromosome Location
Chr. 10: 62035866 - 62109075 on Build Rnor_6.0
UniGene ID
Rn.22361
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181377.3 NP_852042.1 2 793
GenBank mRNA AF532861.1 - 2 548
AY311478.1 - 3 656
BC129084.1 - 2 793 8363864

View Full Product Details