Gene Transcripts

Gene Symbol
Ntsr2
Entrez Gene ID
64636
Gene Name
neurotensin receptor 2
Gene Aliases
Chromosome Location
Chr. 6: 41917094 - 41923780 on Build Rnor_6.0
UniGene ID
Rn.127792
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022695.2 NP_073186.1 4 1275
XM_006239918.2 XP_006239980.1 4 1062
GenBank mRNA X97121.1 - 4 1277

View Full Product Details