Gene Transcripts

Gene Symbol
Akr1a1
Entrez Gene ID
78959
Gene Name
aldo-keto reductase family 1, member A1 (aldehyde reductase)
Gene Aliases
Akr1a4
Chromosome Location
Chr. 5: 135482068 - 135498693 on Build Rnor_6.0
UniGene ID
Rn.835
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031000.3 NP_112262.1 4 301
GenBank mRNA BC059133.1 - 4 580
D10854.1 - 4 301
FQ212523.1 - 4 543
FQ212586.1 - 4 541
FQ212779.1 - 4 576
FQ212919.1 - 3 251
FQ213220.1 - 3 248
FQ213467.1 - 4 576
FQ213483.1 - 4 572
FQ213491.1 - 4 574
FQ213579.1 - 4 543
FQ213776.1 - 4 543
FQ214173.1 - 4 543
FQ214441.1 - 4 320
FQ214642.1 - 558
FQ214700.1 - 4 516
FQ215620.1 - 4 368
FQ215772.1 - 4 543
FQ218383.1 - 4 543
FQ218721.1 - 4 543
FQ218729.1 - 4 543
FQ220055.1 - 2 144
FQ226221.1 - 4 576
FQ228131.1 - 4 576
FQ228132.1 - 4 576
FQ228842.1 - 4 572
FQ229180.1 - 3 215

View Full Product Details