Gene Transcripts

Gene Symbol
Kcnj11
Entrez Gene ID
83535
Gene Name
potassium channel, inwardly rectifying subfamily J, member 11
Gene Aliases
Kir6.2
Chromosome Location
Chr. 1: 102103093 - 102106774 on Build Rnor_6.0
UniGene ID
Rn.3985
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031358.3 NP_112648.2 1 1619
XM_008759421.1 XP_008757643.1 2 1312
GenBank mRNA AB043638.1 - 1 1386

View Full Product Details