Gene Transcripts

Gene Symbol
LYPLA2
Entrez Gene ID
11313
Gene Name
lysophospholipase II
Gene Aliases
APT-2, APT2, DJ886K2.4
Chromosome Location
Chr. 1: 23790789 - 23795539 on Build GRCh38
UniGene ID
Hs.533479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007260.2 NP_009191.1 10 792
XM_005245728.4 XP_005245785.1 10 1264
XM_017000165.1 XP_016855654.1 9 1240
GenBank mRNA AF090423.1 - 10 838
AF098668.1 - 10 792
AK024724.1 - 6 472
BC017034.2 - 10 757 3845764
BC017193.2 - 10 746 3138786

View Full Product Details