Gene Transcripts

Gene Symbol
B4galt6
Entrez Gene ID
65196
Gene Name
UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 6
Gene Aliases
Chromosome Location
Chr. 18: 15462913 - 15525584 on Build Rnor_6.0
UniGene ID
Rn.21126
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031740.1 NP_113928.1 7 1676
GenBank mRNA AF048687.1 - 7 1676

View Full Product Details