Gene Transcripts

Gene Symbol
Cyp27b1
Entrez Gene ID
114700
Gene Name
cytochrome P450, family 27, subfamily b, polypeptide 1
Gene Aliases
Cyp40
Chromosome Location
Chr. 7: 70333150 - 70340006 on Build Rnor_6.0
UniGene ID
Rn.10847
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053763.1 NP_446215.1 9 1509
XM_008765374.1 XP_008763596.1 10 3381
GenBank mRNA AB001992.1 - 9 1509
AF000139.1 - 9 1501

View Full Product Details