Gene Transcripts

Gene Symbol
Stk25
Entrez Gene ID
373542
Gene Name
serine/threonine kinase 25
Gene Aliases
Chromosome Location
Chr. 9: 100767578 - 100779715 on Build Rnor_6.0
UniGene ID
Rn.67557
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_184049.1 NP_908938.1 3, 4 432
GenBank mRNA AY346152.1 - 3, 4 432
BC087092.1 - 3, 4 378 7190997

View Full Product Details