Gene Transcripts

Gene Symbol
Cmtm8
Entrez Gene ID
301045
Gene Name
CKLF-like MARVEL transmembrane domain containing 8
Gene Aliases
Cklfsf8
Chromosome Location
Chr. 8: 122848249 - 122913591 on Build Rnor_6.0
UniGene ID
Rn.103184
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198754.2 NP_942049.1 4 918
XM_006243964.2 XP_006244026.1 4 624
GenBank mRNA AF538323.1 - 4 721
BC088094.1 - 4 918 7325894

View Full Product Details