Gene Transcripts

Gene Symbol
Fgfrl1
Entrez Gene ID
360903
Gene Name
fibroblast growth factor receptor-like 1
Gene Aliases
Fgfr5
Chromosome Location
Chr. 14: 2020110 - 2032169 on Build Rnor_6.0
UniGene ID
Rn.44953
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199114.1 NP_954545.1 4 723
GenBank mRNA AJ536020.1 - 4 723
BC097261.1 - 4 679 7442454

View Full Product Details