Gene Transcripts

Gene Symbol
Cfdp1
Entrez Gene ID
292027
Gene Name
craniofacial development protein 1
Gene Aliases
Chromosome Location
Chr. 19: 43989596 - 44078320 on Build Rnor_6.0
UniGene ID
Rn.128746
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199378.3 NP_955410.1 4 567
GenBank mRNA AB125856.1 - 4 447
BC067246.1 - 4 571 6889499
FQ212332.1 - 601

View Full Product Details