Gene Transcripts

Gene Symbol
Fah
Entrez Gene ID
29383
Gene Name
fumarylacetoacetate hydrolase
Gene Aliases
Chromosome Location
Chr. 1: 146713663 - 146736339 on Build Rnor_6.0
UniGene ID
Rn.9195
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017181.2 NP_058877.1 2, 3 285
GenBank mRNA BC076381.1 - 2, 3 285
FQ219602.1 - 2, 3 243
M77694.1 - 2, 3 207

View Full Product Details